This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02189627_g1
See other EIF6A GE Assays ›
Gene Symbol:
eIF6A
Gene Name:
Translation initiation factor IF6
Gene Aliases:
AT3G55620, emb1624, embryo defective 1624, eukaryotic initiation facor 6A
Chromosome Location:
Chr.3: 20634057 - 20636555 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02189627_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_115420.4 NP_191121.1 5 - 6 536 98
GenBank mRNA AY128351.1 - 5 - 6 359 98
BT009656.1 - 3 - 4 235 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
embryo development ending in seed dormancy
cellular response to abscisic acid stimulus
assembly of large subunit precursor of preribosome
ribosome binding
ribosomal large subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline