This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02189652_g1
See other ROPGEF6 GE Assays ›
Gene Symbol:
ROPGEF6
Gene Name:
ROP (rho of plants) guanine nucleotide exchange factor 6
Gene Aliases:
AT3G55660, ATROPGEF6, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, ROP (rho of plants) guanine nucleotide exchange factor 6
Chromosome Location:
Chr.3: 20648794 - 20651484 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02189652_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339743.1 NP_001325478.1 1 - 2 471 123
NM_115424.3 NP_191125.1 1 - 2 512 123
GenBank mRNA BT020279.1 - 1 - 2 395 123
BT021117.1 - 1 - 2 318 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Process(es)

Rho guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0