This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02189655_gH
See other ROPGEF6 GE Assays ›
Gene Symbol:
ROPGEF6
Gene Name:
ROP (rho of plants) guanine nucleotide exchange factor 6
Gene Aliases:
AT3G55660, ATROPGEF6, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, ROP (rho of plants) guanine nucleotide exchange factor 6
Chromosome Location:
Chr.3: 20648794 - 20651484 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02189655_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339743.1 NP_001325478.1 4 - 5 811 121
NM_115424.3 NP_191125.1 4 - 5 852 121
GenBank mRNA BT020279.1 - 4 - 5 735 121
BT021117.1 - 4 - 5 658 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Process(es)

Rho guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0