This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02191700_g1
See other DND1 GE Assays ›
Gene Symbol:
DND1
Gene Name:
Cyclic nucleotide-regulated ion channel family protein
Gene Aliases:
AT5G15410, ATCNGC2, CNGC2, CYCLIC NUCLEOTIDE GATED CHANNEL 2, CYCLIC NUCLEOTIDE-GATED CHANNEL 2, DEFENSE NO DEATH 1
Chromosome Location:
Chr.5: 5003226 - 5006882 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02191700_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DND1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121545.5 NP_197045.1 5 - 6 1469 92
NM_203054.2 NP_974783.2 6 - 7 1596 92
GenBank mRNA AF067798.1 - 5 - 6 1404 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel ligand-gated ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

defense response
nitric oxide mediated signal transduction
plant-type hypersensitive response
calcium ion import
ion channel activity
intracellular cyclic nucleotide activated cation channel activity
intracellular cAMP activated cation channel activity
inward rectifier potassium channel activity
calcium channel activity
calmodulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0