This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02195189_g1
See other RD21A GE Assays ›
Gene Symbol:
RD21A
Gene Name:
Granulin repeat cysteine protease family protein
Gene Aliases:
AT1G47128, F2G19.31, F2G19_31, RD21, RESPONSIVE TO DEHYDRATION 21A, responsive to dehydration 21, responsive to dehydration 21A
Chromosome Location:
Chr.1: 17282749 - 17285749 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02195189_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RD21A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_103612.3 NP_564497.1 2 - 3 848 88
GenBank mRNA AY072130.1 - 2 - 3 768 88
AY133781.1 - 2 - 3 708 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
response to water deprivation
defense response to fungus
proteolysis involved in cellular protein catabolic process
cysteine-type endopeptidase activity
protein binding
peptidase activity
cysteine-type peptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0