This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02198705_g1
See other PBS3 GE Assays ›
Gene Symbol:
PBS3
Gene Name:
Auxin-responsive GH3 family protein
Gene Aliases:
AT5G13320, AVRPPHB SUSCEPTIBLE 3, AtGH3.12, GDG1, GH3-LIKE DEFENSE GENE 1, GH3.12, GRETCHEN HAGEN 3.12, HOPW1-1-INTERACTING 3, T22N19.5, WIN3
Chromosome Location:
Chr.5: 4267456 - 4271064 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02198705_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PBS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001343268.1 NP_001330075.1 2 - 3 639 91
NM_001343269.1 NP_001330076.1 1 - 2 652 91
NM_121335.4 NP_196836.1 2 - 3 429 91
GenBank mRNA EU214910.1 - 1 - 2 282 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

defense response
defense response to bacterium, incompatible interaction
salicylic acid mediated signaling pathway
regulation of systemic acquired resistance
detection of fungus
benzoate metabolic process
positive regulation of plant-type hypersensitive response
defense response to bacterium
interspecies interaction between organisms
cellular response to hypoxia
acid-amino acid ligase activity
4-aminobenzoate amino acid synthetase activity
benzoate amino acid synthetase activity
vanillate amino acid synthetase activity
4-hydroxybenzoate amino acid synthetase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline