This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02200913_g1
See other XPO1A GE Assays ›
Gene Symbol:
XPO1A
Gene Name:
exportin 1A
Gene Aliases:
ARABIDOPSIS THALIANA EXPORTIN 1, AT5G17020, ATCRM1, ATXPO1, EXPORTIN 1, F2K13.170, F2K13_170, HEAT-INTOLERANT 2, HIT2, XPO1, exportin 1A
Chromosome Location:
Chr.5: 5594632 - 5602969 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02200913_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPO1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203395.1 NP_001190324.1 15 - 16 1683 75
NM_121708.3 NP_197204.1 16 - 17 1820 75
GenBank mRNA AY056126.1 - 16 - 17 1716 75
AY096633.1 - 16 - 17 1548 75
Y18469.1 - 1643 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit export from nucleus
ribosomal small subunit export from nucleus
protein export from nucleus
response to heat
embryo sac development
pollen development
pollen germination
pollen tube growth
regulation of protein export from nucleus
nuclear export signal receptor activity
protein binding
Ran GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline