Assay Details
| Gene Symbol: AGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128854.4 | NP_180853.2 | 22 - 23 | 2686 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGO6| Gene Symbol: AGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128854.4 | NP_180853.2 | 22 - 23 | 2686 | 59 | |
Set Membership: |
|