This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02204936_g1
See other ABCG12 GE Assays ›
Gene Symbol:
ABCG12
Gene Name:
ABC-2 type transporter family protein
Gene Aliases:
ABC TRANSPORTER FROM THE ABCG SUBFAMILY 12, ARABIDOPSIS THALIANA WHITE-BROWN COMPLEX 12, AT1G51500, ATP-binding cassette G12, ATWBC12, AtABCG12, CER5, D3, ECERIFERUM 5, F5D21.6, F5D21_6, WBC12, WHITE-BROWN COMPLEX 12
Chromosome Location:
Chr.1: 19097712 - 19101265 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02204936_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104028.5 NP_175561.1 5 - 6 1323 66
GenBank mRNA AF412079.1 - 5 - 6 1125 66
AY093162.1 - 5 - 6 1115 66
BT008427.1 - 5 - 6 1030 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

response to salt stress
response to abscisic acid
wax biosynthetic process
defense response to virus
transmembrane transport
protein binding
efflux transmembrane transporter activity
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2hh2p:80/100.66.78.76:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0