This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02204938_g1
See other ABCG12 GE Assays ›
Gene Symbol:
ABCG12
Gene Name:
ABC-2 type transporter family protein
Gene Aliases:
ABC TRANSPORTER FROM THE ABCG SUBFAMILY 12, ARABIDOPSIS THALIANA WHITE-BROWN COMPLEX 12, AT1G51500, ATP-binding cassette G12, ATWBC12, AtABCG12, CER5, D3, ECERIFERUM 5, F5D21.6, F5D21_6, WBC12, WHITE-BROWN COMPLEX 12
Chromosome Location:
Chr.1: 19097712 - 19101265 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02204938_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104028.5 NP_175561.1 7 - 8 1988 90
GenBank mRNA AF412079.1 - 7 - 8 1790 90
AY093162.1 - 7 - 8 1780 90
BT008427.1 - 7 - 8 1695 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

response to salt stress
response to abscisic acid
wax biosynthetic process
defense response to virus
transmembrane transport
protein binding
efflux transmembrane transporter activity
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline