This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02208896_gH
See other RBP-DR1 GE Assays ›
Gene Symbol:
RBP-DR1
Gene Name:
RNA-binding protein-defense related 1
Gene Aliases:
AT4G03110, AtBRN1, AtRBP-DR1, BRN1, Bruno-like 1, F4C21.3, RNA-binding protein-defense related 1
Chromosome Location:
Chr.4: 1376306 - 1379747 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02208896_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBP-DR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340422.1 NP_001329301.1 3 - 4 564 118
NM_116545.4 NP_567249.1 3 - 4 752 118
NM_178963.3 NP_849294.1 3 - 4 784 118
GenBank mRNA AK316961.1 - 3 - 4 497 118
AY065261.1 - 3 - 4 411 118
AY084669.1 - 3 - 4 440 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
plant-type hypersensitive response
positive regulation of salicylic acid mediated signaling pathway
regulation of photoperiodism, flowering
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline