This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02208959_g1
See other FH GE Assays ›
Gene Symbol:
FH
Gene Name:
frataxin-like protein
Gene Aliases:
AT4G03240, ATFH, F4C21.17, F4C21_17, frataxin homolog
Chromosome Location:
Chr.4: 1423238 - 1424772 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02208959_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_116562.4 NP_192233.2 1 - 2 234 72
GenBank mRNA AK176448.1 - 1 - 2 232 72
AK176831.1 - 1 - 2 234 72
AY649366.1 - 1 - 2 220 72
BT014964.1 - 1 - 2 220 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

cellular iron ion homeostasis
response to oxidative stress
mitochondrion organization
aerobic respiration
embryo development ending in seed dormancy
iron incorporation into metallo-sulfur cluster
response to hydrogen peroxide
regulation of iron-sulfur cluster assembly
ferroxidase activity
ferrous iron binding
ferric iron binding
iron chaperone activity
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0