This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02209566_mH
See other MSRB3 GE Assays ›
Gene Symbol:
MSRB3
Gene Name:
methionine sulfoxide reductase B3
Gene Aliases:
AT4G04800, ATMSRB3, T4B21.6, T4B21_6, methionine sulfoxide reductase B3
Chromosome Location:
Chr.4: 2439520 - 2441205 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02209566_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSRB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_116718.3 NP_567271.1 1 - 2 269 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cold acclimation
protein repair
cellular response to oxidative stress
oxidation-reduction process
peptide-methionine (S)-S-oxide reductase activity
peptide-methionine (R)-S-oxide reductase activity
methionine-R-sulfoxide reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline