This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02212239_g1
See other AT1G54240 GE Assays ›
Gene Symbol:
AT1G54240
Gene Name:
winged-helix DNA-binding transcription factor family protein
Gene Aliases:
AT1G54240, F20D21.6, F20D21_6
Chromosome Location:
Chr.1: 20252718 - 20254708 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02212239_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G54240
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104302.4 NP_175826.2 6 - 7 516 125
GenBank mRNA DQ446358.1 - 5 - 6 339 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
regulation of transcription, DNA-templated
nucleosome positioning
chromosome condensation
negative regulation of chromatin silencing
negative regulation of DNA recombination
double-stranded DNA binding
nucleosomal DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0