This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02215586_g1
See other MLO6 GE Assays ›
Gene Symbol:
MLO6
Gene Name:
Seven transmembrane MLO family protein
Gene Aliases:
AT1G61560, ATMLO6, MILDEW RESISTANCE LOCUS O 6, T25B24.9, T25B24_9
Chromosome Location:
Chr.1: 22708468 - 22712289 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02215586_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLO6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001333983.1 NP_001322619.1 1 - 2 353 126
NM_001333984.1 NP_001319285.1 1 - 2 298 126
NM_001333985.1 NP_001322617.1 1 - 2 377 126
NM_001333986.1 NP_001322615.1 1 - 2 377 126
NM_001333987.1 NP_001322616.1 1 - 2 377 126
NM_001333988.1 NP_001322614.1 1 - 2 377 126
NM_001333989.1 NP_001322618.1 1 - 2 353 126
NM_104836.2 NP_176350.1 1 - 2 377 126
GenBank mRNA AF369567.2 - 1 - 2 132 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s)

defense response to fungus, incompatible interaction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0