This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02215590_g1
See other MLO6 GE Assays ›
Gene Symbol:
MLO6
Gene Name:
Seven transmembrane MLO family protein
Gene Aliases:
AT1G61560, ATMLO6, MILDEW RESISTANCE LOCUS O 6, T25B24.9, T25B24_9
Chromosome Location:
Chr.1: 22708468 - 22712289 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02215590_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLO6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198352.1 NP_001185281.1 5 - 6 628 69
NM_001198354.1 NP_001185283.1 5 - 6 630 69
NM_001333983.1 NP_001322619.1 5 - 6 848 69
NM_001333984.1 NP_001319285.1 5 - 6 793 69
NM_001333985.1 NP_001322617.1 5 - 6 872 69
NM_001333986.1 NP_001322615.1 5 - 6 872 69
NM_001333987.1 NP_001322616.1 5 - 6 872 69
NM_001333988.1 NP_001322614.1 5 - 6 872 69
NM_001333989.1 NP_001322618.1 5 - 6 848 69
NM_104836.2 NP_176350.1 5 - 6 872 69
GenBank mRNA AF369567.2 - 5 - 6 627 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s)

defense response to fungus, incompatible interaction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0