This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02215710_g1
See other GPT2 GE Assays ›
Gene Symbol:
GPT2
Gene Name:
glucose-6-phosphate/phosphate translocator 2
Gene Aliases:
ARABIDOPSIS GLUCOSE-6-PHOSPHATE/PHOSPHATE TRANSLOCATOR 2, AT1G61800, ATGPT2, GLUCOSE6-PHOSPHATE/PHOSPHATE TRANSPORTER 2, T13M11.18, T13M11_18, glucose-6-phosphate/phosphate translocator 2
Chromosome Location:
Chr.1: 22824414 - 22826902 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02215710_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334001.1 NP_001320539.1 2 - 3 829 75
NM_104862.4 NP_564785.1 2 - 3 829 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

gamete generation
response to nematode
photosynthetic acclimation
response to sucrose
response to glucose
regulation of photosynthesis
phosphoglycerate transport
phosphoenolpyruvate transport
glucose-6-phosphate transport
photosynthesis
triose phosphate transmembrane transport
transmembrane transport
response to karrikin
inorganic phosphate transmembrane transporter activity
phosphoglycerate transmembrane transporter activity
glucose-6-phosphate transmembrane transporter activity
antiporter activity
transmembrane transporter activity
triose-phosphate transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0