This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02223397_g1
See other DSEL GE Assays ›
Gene Symbol:
DSEL
Gene Name:
alpha/beta-Hydrolases superfamily protein
Gene Aliases:
AT4G18550, Arabidopsis thaliana DAD1-like seeding establishment-related lipase, AtDSEL, DAD1-like seeding establishment-related lipase, F28J12.210, F28J12_210
Chromosome Location:
Chr.4: 10224661 - 10227140 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02223397_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001341292.1 NP_001328832.1 3 - 4 1140 93
NM_117969.3 NP_193590.1 3 - 4 862 93
GenBank mRNA BT030056.1 - 3 - 4 862 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
negative regulation of seed germination
lipid storage
diacylglycerol catabolic process
monoacylglycerol catabolic process
phosphatidylcholine 1-acylhydrolase activity
acylglycerol lipase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0