This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02226560_mH
See other RPL34 GE Assays ›
Gene Symbol:
RPL34
Gene Name:
ribosomal protein L34
Gene Aliases:
AT1G69620, F24J1.23, ribosomal protein L34
Chromosome Location:
Chr.1: 26189775 - 26191404 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02226560_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105630.4 NP_177120.1 2 - 3 275 97
GenBank mRNA AF324703.1 - 2 - 3 212 97
AF327531.1 - 2 - 3 211 97
AF349526.1 - 2 - 3 150 97
AF446885.1 - 2 - 3 150 97
AY052720.1 - 2 - 3 212 97
AY085544.1 - 2 - 3 212 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
ribosome biogenesis
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0