This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02229821_g1
See other ASF1B GE Assays ›
Gene Symbol:
ASF1B
Gene Name:
anti- silencing function 1b
Gene Aliases:
AT5G38110, F16F17.110, F16F17_110, SGA01, SGA1, anti- silencing function 1b
Chromosome Location:
Chr.5: 15208358 - 15210335 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02229821_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASF1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123171.4 NP_198627.1 2 - 3 510 73
GenBank mRNA AB078340.1 - 2 - 3 375 73
AY072420.1 - 2 - 3 394 73
AY086448.1 - 2 - 3 396 73
AY114716.1 - 2 - 3 225 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
regulation of cell size
DNA mediated transformation
trichome branching
mitotic cell size control checkpoint
cell division
protein binding
histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0