This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02233431_g1
See other ALD1 GE Assays ›
Gene Symbol:
ALD1
Gene Name:
AGD2-like defense response protein 1
Gene Aliases:
AGD2-like defense response protein 1, AT2G13810, AtALD1, EDTS5, F17L24.14, eds two suppressor 5
Chromosome Location:
Chr.2: 5768417 - 5772266 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02233431_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_126957.2 NP_565359.1 3 - 4 241 87
GenBank mRNA AY057526.1 - 3 - 4 212 87
AY143898.1 - 3 - 4 169 87
AY518702.1 - 3 - 4 169 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

systemic acquired resistance
systemic acquired resistance, salicylic acid mediated signaling pathway
leaf senescence
defense response to bacterium
L-pipecolic acid biosynthetic process
transaminase activity
pyridoxal phosphate binding
L-lysine alpha-aminotransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline