Assay Details
| Gene Symbol: MAP1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101198.4 | NP_172785.1 | 5 - 6 | 779 | 115 | |
| NM_202086.1 | NP_973815.1 | 5 - 6 | 712 | 115 | ||
| GenBank mRNA | BT000449.1 | - | 5 - 6 | 712 | 115 | |
| BT001213.1 | - | 5 - 6 | 669 | 115 | ||
More Information
Set Membership: |
|

MAP1C
Genomic Map