This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02234123_g1
See other MAP1C GE Assays ›
Gene Symbol:
MAP1C
Gene Name:
methionine aminopeptidase 1B
Gene Aliases:
AT1G13270, MAP1B, METHIONINE AMINOPEPTIDASE 1B, methionine aminopeptidase 1B
Chromosome Location:
Chr.1: 4544889 - 4547373 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02234123_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAP1C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_101198.4 NP_172785.1 6 - 7 849 74
NM_202086.1 NP_973815.1 6 - 7 782 74
GenBank mRNA BT000449.1 - 6 - 7 782 74
BT001213.1 - 6 - 7 739 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid modification
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline