This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02236224_g1
See other SFR2 GE Assays ›
Gene Symbol:
SFR2
Gene Name:
Glycosyl hydrolase superfamily protein
Gene Aliases:
AT3G06510, ATSFR2, F5E6.16, F5E6_16, SENSITIVE TO FREEZING 2
Chromosome Location:
Chr.3: 2016246 - 2020103 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02236224_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SFR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001125119.2 NP_001118591.1 2 - 3 564 74
NM_111527.3 NP_566285.1 2 - 3 647 74
GenBank mRNA AJ491320.1 - 2 - 3 563 74
AY059889.1 - 2 - 3 488 74
BT002100.1 - 2 - 3 443 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
response to cold
response to freezing
galactosyltransferase activity
beta-glucosidase activity
transferase activity, transferring glycosyl groups
hydrolase activity, acting on glycosyl bonds
galactolipid galactosyltransferase activity
cellobiose glucosidase activity
beta,beta digalactosyldiacylglycerol galactosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0