This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02237001_g1
See other ADT2 GE Assays ›
Gene Symbol:
ADT2
Gene Name:
arogenate dehydratase 2
Gene Aliases:
AT3G07630, Arabidopsis thaliana arogenate dehydratase 2, AtADT2, arogenate dehydratase 2
Chromosome Location:
Chr.3: 2435408 - 2437989 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02237001_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337726.1 NP_001325505.1 5 - 6 722 84
NM_111642.3 NP_187420.1 5 - 6 740 84
NM_202520.2 NP_974249.1 5 - 6 703 84
GenBank mRNA AY050813.1 - 5 - 6 698 84
AY113967.1 - 5 - 6 691 84
DQ411465.1 - 5 - 6 695 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

L-phenylalanine biosynthetic process
prephenate dehydratase activity
arogenate dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline