This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02243079_g1
See other AT4G31270 GE Assays ›
Gene Symbol:
AT4G31270
Gene Name:
sequence-specific DNA binding transcription factor
Gene Aliases:
AT4G31270, F8F16.90, F8F16_90
Chromosome Location:
Chr.4: 15183188 - 15184961 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02243079_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G31270
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119276.3 NP_194855.2 2 - 3 545 73
GenBank mRNA AK118674.1 - 2 - 3 545 73
BT005287.1 - 1 - 2 453 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of chromatin silencing
DNA demethylation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ln7zh:80/100.66.79.20:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0