This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02244755_g1
See other MCA1 GE Assays ›
Gene Symbol:
MCA1
Gene Name:
PLAC8 family protein
Gene Aliases:
AT4G35920, T19K4.50, mid1-complementing activity 1
Chromosome Location:
Chr.4: 17011788 - 17015493 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02244755_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001342380.1 NP_001329715.1 8 - 9 1644 74
NM_001342381.1 NP_001329716.1 8 - 9 1532 74
NM_119759.4 NP_195317.2 8 - 9 1872 74
NM_179172.3 NP_849503.1 8 - 9 1566 74
NM_179173.4 NP_849504.2 8 - 9 1793 74
GenBank mRNA AB196960.1 - 8 - 9 1600 74
AK316845.1 - 8 - 9 1567 74
AY136293.1 - 8 - 9 1624 74
BT003430.1 - 6 - 7 1216 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
osmosensory signaling pathway
carbohydrate homeostasis
calcium ion transmembrane transport
cellular response to mechanical stimulus
calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0