This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02246259_g1
See other ABCG26 GE Assays ›
Gene Symbol:
ABCG26
Gene Name:
ABC-2 type transporter family protein
Gene Aliases:
AT3G13220, ATP BINDING CASSETTE G26, ATP-binding cassette G26, AtABCG26, MSR02, WBC27
Chromosome Location:
Chr.3: 4247675 - 4250703 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02246259_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_112162.3 NP_187928.2 2 - 3 490 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

pollen development
pollen maturation
pollen exine formation
polyketide metabolic process
transmembrane transport
sporopollenin biosynthetic process
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0