This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02249173_g1
See other EIF3K GE Assays ›
Gene Symbol:
EIF3K
Gene Name:
eukaryotic translation initiation factor 3K
Gene Aliases:
AT4G33250, ATTIF3K1, F17M5.10, F17M5_10, TIF3K1, eukaryotic translation initiation factor 3K
Chromosome Location:
Chr.4: 16038651 - 16040808 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02249173_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119479.4 NP_195051.1 5 - 6 593 72
GenBank mRNA AF291713.1 - 5 - 6 461 72
AY037191.1 - 5 - 6 486 72
AY050928.1 - 5 - 6 486 72
AY091164.1 - 5 - 6 402 72
AY091670.1 - 5 - 6 402 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of translational initiation
translation initiation factor activity
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw9fd:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0