This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02249263_g1
See other ABCI10 GE Assays ›
Gene Symbol:
ABCI10
Gene Name:
ABC transporter family protein
Gene Aliases:
AT4G33460, ATNAP13, ATP-binding cassette I10, EMB2751, F17M5.220, F17M5_220, embryo defective 2751
Chromosome Location:
Chr.4: 16098070 - 16100440 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02249263_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCI10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119500.6 NP_195072.2 4 - 5 924 94
GenBank mRNA AK220760.1 - 4 - 5 750 94
AY142518.1 - 4 - 5 597 94
BT010649.1 - 4 - 5 597 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

chloroplast organization
embryo development ending in seed dormancy
transition metal ion homeostasis
protein binding
ATPase activity
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0