This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02250631_g1
See other BBD1 GE Assays ›
Gene Symbol:
BBD1
Gene Name:
bifunctional nuclease in basal defense response 1
Gene Aliases:
AT1G75380, ATBBD1, F1B16.9, F1B16_9, bifunctional nuclease in basal defense response 1
Chromosome Location:
Chr.1: 28281195 - 28284168 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02250631_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BBD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334685.1 NP_001322708.1 7 - 8 1007 119
NM_106192.3 NP_177671.1 7 - 8 884 119
NM_179559.3 NP_849890.1 7 - 8 1007 119
NM_179560.3 NP_849891.1 7 - 8 871 119
GenBank mRNA AY054232.1 - 7 - 8 876 119
AY066045.1 - 6 - 7 712 119
AY087417.1 - 7 - 8 881 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of histone deacetylation
negative regulation of transcription, DNA-templated
defense response to fungus
nuclease activity
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline