This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02251778_g1
See other NLP1 GE Assays ›
Gene Symbol:
NLP1
Gene Name:
nitrilase-like protein 1
Gene Aliases:
AT2G27450, ATNLP1, CPA, F10A12.13, F10A12_13, N-CARBAMOYLPUTRESCINE AMIDOHYDROLASE, nitrilase-like protein 1
Chromosome Location:
Chr.2: 11737374 - 11739690 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02251778_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NLP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001336116.1 NP_001324215.1 7 - 8 1064 75
NM_128305.2 NP_565650.1 8 - 9 847 75
NM_179770.1 NP_850101.1 8 - 9 836 75
GenBank mRNA AK317300.1 - 8 - 9 767 75
AY072113.1 - 8 - 9 836 75
AY086056.1 - 8 - 9 847 75
AY122963.1 - 8 - 9 815 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nitrogen compound metabolic process
putrescine biosynthetic process
putrescine biosynthetic process from arginine
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in linear amides
N-carbamoylputrescine amidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline