This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02255533_gH
See other CAT2 GE Assays ›
Gene Symbol:
CAT2
Gene Name:
catalase 2
Gene Aliases:
AT4G35090, CATALASE, T12J5.2, catalase 2
Chromosome Location:
Chr.4: 16700312 - 16703504 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02255533_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001342324.1 NP_001328611.1 6 - 7 1946 70
NM_119675.4 NP_195235.1 7 - 8 1704 70
GenBank mRNA AY074301.1 - 7 - 8 1487 70
AY113854.1 - 6 - 7 1415 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
cellular response to nitrogen starvation
cell death
response to cold
response to light stimulus
photoperiodism
cellular response to sulfate starvation
cellular response to phosphate starvation
response to hydrogen peroxide
hydrogen peroxide catabolic process
cell redox homeostasis
catalase activity
protein binding
heme binding
cobalt ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline