This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02256405_g1
See other AT4G36590 GE Assays ›
Gene Symbol:
AT4G36590
Gene Name:
MADS-box transcription factor family protein
Gene Aliases:
AP22.88, AP22_88, AT4G36590
Chromosome Location:
Chr.4: 17260937 - 17262227 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02256405_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G36590
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119822.1 NP_195377.1 1 - 2 371 87
GenBank mRNA AY086178.1 - 1 - 2 371 87
AY141234.1 - 1 - 2 333 87
DQ446902.1 - 1 - 2 333 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

MADS box transcription factor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of transcription from RNA polymerase II promoter
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0