This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02257816_g1
See other BZIP16 GE Assays ›
Gene Symbol:
bZIP16
Gene Name:
basic region/leucine zipper transcription factor 16
Gene Aliases:
AT2G35530, AtbZIP16, T32F12.9, T32F12_9, basic region/leucine zipper transcription factor 16
Chromosome Location:
Chr.2: 14922836 - 14926609 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02257816_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: bZIP16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_179917.4 NP_850248.2 10 - 11 1108 80
GenBank mRNA AB493577.1 - 9 - 10 909 80
BT022055.1 - 10 - 11 1018 80
BT026441.1 - 9 - 10 909 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
identical protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline