Assay Details
| Gene Symbol: HB22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179935.2 | NP_850266.1 | 2 - 3 | 601 | 151 | |
| GenBank mRNA | DQ056569.1 | - | 2 - 3 | 485 | 151 | |
More Information
Set Membership: |
|

HB22
Genomic Map