This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02258629_g1
See other ROPGEF2 GE Assays ›
Gene Symbol:
ROPGEF2
Gene Name:
RHO guanyl-nucleotide exchange factor 2
Gene Aliases:
AT1G01700, ATROPGEF2, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, RHO guanyl-nucleotide exchange factor 2, ROP (rho of plants) guanine nucleotide exchange factor 2, T1N6.8, T1N6_8
Chromosome Location:
Chr.1: 259307 - 261667 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02258629_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331294.1 NP_001321886.1 3 - 4 812 129
NM_001331296.1 NP_001321884.1 3 - 4 760 129
NM_001331297.1 NP_001321885.1 3 - 4 812 129
NM_100052.2 NP_171676.1 3 - 4 812 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Process(es)

Rho guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0