This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02259100_m1
See other GPAT2 GE Assays ›
Gene Symbol:
GPAT2
Gene Name:
glycerol-3-phosphate acyltransferase 2
Gene Aliases:
ARABIDOPSIS THALIANA GLYCEROL-3-PHOSPHATE ACYLTRANSFERASE 2, AT1G02390, ATGPAT2, GLYCEROL-3-PHOSPHATE sn-2-ACYLTRANSFERASE 2, T6A9.8, T6A9_8, glycerol-3-phosphate acyltransferase 2
Chromosome Location:
Chr.1: 480650 - 483373 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02259100_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_100120.3 NP_563651.1 1 - 2 974 82
GenBank mRNA AF419560.1 - 1 - 2 772 82
AY097339.1 - 1 - 2 703 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

cutin biosynthetic process
CDP-diacylglycerol biosynthetic process
glycerol-3-phosphate O-acyltransferase activity
phosphatase activity
glycerol-3-phosphate 2-O-acyltransferase activity
sn-1-glycerol-3-phosphate C16:0-DCA-CoA acyl transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0