This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02262796_g1
See other WRKY12 GE Assays ›
Gene Symbol:
WRKY12
Gene Name:
WRKY family transcription factor
Gene Aliases:
AT2G44745, AtWRKY12, WRKY DNA-binding protein 12
Chromosome Location:
Chr.2: 18447203 - 18449093 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02262796_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WRKY12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130039.3 NP_566025.2 3 - 4 581 66
GenBank mRNA AF404857.1 - 3 - 4 492 66
BT028992.1 - 3 - 4 492 66
DQ446622.1 - 3 - 4 492 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0