This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02263493_g1
See other ROPGEF4 GE Assays ›
Gene Symbol:
ROPGEF4
Gene Name:
RHO guanyl-nucleotide exchange factor 4
Gene Aliases:
AT2G45890, ATROPGEF4, F4I18.13, GEF4, RHO GUANYL-NUCLEOTIDE EXCHANGE FACTOR 4, RHO guanyl-nucleotide exchange factor 4, RHS11, ROOT HAIR SPECIFIC 11, ROP (rho of plants) guanine nucleotide exchange factor 4, guanine exchange factor 4
Chromosome Location:
Chr.2: 18883377 - 18885729 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02263493_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130152.3 NP_182113.2 6 - 7 1430 76
GenBank mRNA AK229390.1 - 6 - 7 1334 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

plant-type cell wall organization
defense response to fungus, incompatible interaction
root hair elongation
Rho guanyl-nucleotide exchange factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0