This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02266917_g1
See other MLO4 GE Assays ›
Gene Symbol:
MLO4
Gene Name:
Seven transmembrane MLO family protein
Gene Aliases:
AT1G11000, ATMLO4, MILDEW RESISTANCE LOCUS O 4, T19D16.26, T19D16_26
Chromosome Location:
Chr.1: 3671756 - 3676133 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02266917_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331947.1 NP_001320915.1 3 - 4 540 84
NM_001331948.1 NP_001320916.1 4 - 5 532 84
NM_100975.3 NP_563882.1 3 - 4 548 84
GenBank mRNA AF369565.1 - 3 - 4 455 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0