This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02267409_g1
See other GNC GE Assays ›
Gene Symbol:
GNC
Gene Name:
GATA type zinc finger transcription factor family protein
Gene Aliases:
AT5G56860, GATA, GATA TRANSCRIPTION FACTOR 21, GATA21, MPI10.2, MPI10_2, carbon metabolism-involved, nitrate-inducible
Chromosome Location:
Chr.5: 22989216 - 22991530 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02267409_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125069.3 NP_200497.1 1 - 2 456 66
GenBank mRNA BT025970.1 - 1 - 2 277 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
circadian rhythm
response to light stimulus
chloroplast organization
response to auxin
response to cytokinin
cytokinin-activated signaling pathway
response to gibberellin
gibberellic acid mediated signaling pathway
negative regulation of flower development
negative regulation of gibberellic acid mediated signaling pathway
regulation of seed germination
chloroplast elongation
negative regulation of seed germination
glucose mediated signaling pathway
regulation of chlorophyll biosynthetic process
regulation of gene expression
regulation of carbohydrate utilization
regulation of nitrogen compound metabolic process
regulation of seed development
plant organ senescence
positive regulation of chlorophyll biosynthetic process
regulation of photoperiodism, flowering
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline