This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02268167_g1
See other MSI1 GE Assays ›
Gene Symbol:
MSI1
Gene Name:
Transducin/WD40 repeat-like superfamily protein
Gene Aliases:
ARABIDOPSIS MULTICOPY SUPRESSOR OF IRA1, AT5G58230, ATMSI1, MATERNAL EFFECT EMBRYO ARREST 70, MCK7.10, MCK7_10, MEE70, MULTICOPY SUPRESSOR OF IRA1
Chromosome Location:
Chr.5: 23556012 - 23558245 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02268167_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125208.3 NP_200631.1 5 - 6 1143 70
GenBank mRNA AF016846.1 - 5 - 6 1118 70
AY059874.1 - 5 - 6 1123 70
AY114646.1 - 5 - 6 1043 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of gene expression by genetic imprinting
multicellular organism development
pollen development
embryo development ending in seed dormancy
regulation of flower development
trichome differentiation
seed coat development
heterochromatin assembly
positive regulation of cell cycle
seed development
leaf development
heterochromatin organization
regulation of genetic imprinting
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0