This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02268187_g1
See other ABCB25 GE Assays ›
Gene Symbol:
ABCB25
Gene Name:
ABC transporter of the mitochondrion 3
Gene Aliases:
ABC transporter of the mitochondrion 3, ARABIDOPSIS THALIANA ABC TRANSPORTER OF THE MITOCHONDRION 3, AT5G58270, ATATM3, ATM3, ATP-binding cassette B25, HALF-MOLECULE ABC TRANSPORTER ATM3, MCK7.14, MCK7_14, STA1, STARIK 1
Chromosome Location:
Chr.5: 23561890 - 23567244 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02268187_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCB25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125212.3 NP_200635.1 6 - 7 1323 117
GenBank mRNA AF287699.1 - 6 - 7 1045 117
AF360334.1 - 6 - 7 1068 117
AY142686.1 - 6 - 7 1045 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

Mo-molybdopterin cofactor biosynthetic process
cellular iron ion homeostasis
pollen development
chloroplast organization
response to lead ion
regulation of chlorophyll biosynthetic process
regulation of multicellular organism growth
response to cadmium ion
root development
regulation of catalytic activity
chromosome organization
transmembrane transport
ATPase activity
transmembrane transporter activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0