This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02269646_g1
See other ABCG28 GE Assays ›
Gene Symbol:
ABCG28
Gene Name:
ABC transporter family protein
Gene Aliases:
AT5G60740, ATP-binding cassette G28, MUP24.11, MUP24_11
Chromosome Location:
Chr.5: 24425455 - 24430413 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02269646_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125467.5 NP_200882.4 4 - 5 787 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

pollen tube growth
polyamine transport
transmembrane transport
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline