Assay Details
| Gene Symbol: AT2G26230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128180.5 | NP_180191.1 | 2 - 3 | 406 | 72 | |
| GenBank mRNA | AY050318.1 | - | 2 - 3 | 309 | 72 | |
| AY087805.1 | - | 2 - 3 | 359 | 72 | ||
| AY101530.1 | - | 1 - 2 | 209 | 72 | ||
More Information
Set Membership: |
|

AT2G26230
Genomic Map