This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02283583_gH
See other MLO2 GE Assays ›
Gene Symbol:
MLO2
Gene Name:
Seven transmembrane MLO family protein
Gene Aliases:
AT1G11310, ATMLO2, MILDEW RESISTANCE LOCUS O 2, PMR2, POWDERY MILDEW RESISTANT 2, T28P6.4, T28P6_4
Chromosome Location:
Chr.1: 3800500 - 3804233 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02283583_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001123796.1 NP_001117268.1 10 - 11 1466 76
NM_001331977.1 NP_001320358.1 9 - 10 1164 76
NM_101004.4 NP_172598.1 10 - 11 1514 76
GenBank mRNA AF369563.1 - 10 - 11 1214 76
AK226449.1 - 10 - 11 1467 76
AY086586.1 - 10 - 11 1425 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s)

response to fungus
defense response to fungus, incompatible interaction
negative regulation of defense response

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0