This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02285281_g1
See other FLP GE Assays ›
Gene Symbol:
FLP
Gene Name:
Duplicated homeodomain-like superfamily protein
Gene Aliases:
AT1G14350, AtMYB124, F14L17.12, F14L17_12, FOUR LIPS, MYB124, myb domain protein 124
Chromosome Location:
Chr.1: 4907887 - 4911168 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02285281_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FLP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084065.3 NP_001077534.1 7 - 8 909 92
NM_001332112.1 NP_001321792.1 6 - 7 877 92
NM_001332113.1 NP_001321793.1 5 - 6 666 92
NM_101302.3 NP_563948.1 6 - 7 674 92
GenBank mRNA AF371982.2 - 6 - 7 724 92
BT026126.1 - 6 - 7 496 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
embryo sac development
megasporogenesis
response to gravity
response to auxin
response to abscisic acid
auxin polar transport
guard cell differentiation
guard mother cell cytokinesis
stomatal complex formation
guard mother cell differentiation
regulation of DNA endoreduplication
multicellular organismal water homeostasis
positive regulation of histone H3-K27 methylation
primary root development
leaf pavement cell development
positive regulation of response to salt stress
positive regulation of lateral root development
positive regulation of response to water deprivation
regulation of cell cycle G1/S phase transition
regulation of stomatal complex patterning
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0