This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02285463_g1
See other SDS GE Assays ›
Gene Symbol:
SDS
Gene Name:
Cyclin family protein
Gene Aliases:
AT1G14750, F10B6.15, F10B6_15, SOLO DANCERS
Chromosome Location:
Chr.1: 5079226 - 5082536 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02285463_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084070.2 NP_001077539.1 3 - 4 685 65
NM_001332135.1 NP_001322548.1 2 - 3 1302 65
NM_001332136.1 NP_001322547.1 3 - 4 685 65
NM_101344.3 NP_172928.2 2 - 3 1302 65
GenBank mRNA AJ457977.1 - 2 - 3 1286 65
DQ446250.1 - 3 - 4 685 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
double-strand break repair via homologous recombination
mitotic cell cycle phase transition
chiasma assembly
cyclin-dependent protein serine/threonine kinase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0