This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02286435_m1
See other RPS9 GE Assays ›
Gene Symbol:
RPS9
Gene Name:
ribosomal protein S9
Gene Aliases:
AT1G74970, F25A4.6, F25A4_6, RIBOSOMAL PROTEIN S9, TWN3, ribosomal protein S9
Chromosome Location:
Chr.1: 28157386 - 28159317 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02286435_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106155.4 NP_177635.1 1 - 2 448 71
GenBank mRNA AB022676.1 - 1 - 2 409 71
AF344318.1 - 1 - 2 333 71
AK175127.1 - 1 - 2 400 71
AK175501.1 - 1 - 2 383 71
AK175562.1 - 1 - 2 383 71
AK175742.1 - 1 - 2 422 71
AK175827.1 - 1 - 2 382 71
AK175895.1 - 1 - 2 382 71
AK175968.1 - 1 - 2 382 71
AK176368.1 - 1 - 2 400 71
AK176696.1 - 1 - 2 383 71
AK176754.1 - 1 - 2 400 71
AK176758.1 - 1 - 2 383 71
AK176773.1 - 1 - 2 396 71
AK176790.1 - 1 - 2 385 71
AK176797.1 - 1 - 2 433 71
AK176858.1 - 1 - 2 383 71
AY045600.1 - 1 - 2 382 71
AY045856.1 - 1 - 2 400 71
AY086419.1 - 1 - 2 382 71
AY091382.1 - 1 - 2 333 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
RNA binding
mRNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0